Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Benzoate, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003439 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-439 500 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-439 Supplier Avantor Supplier No. Q13907

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 93-58-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key QPJVMBTYPHYUOC-UHFFFAOYSA-N
Synonym methylbenzoate, benzoic acid, methyl ester, clorius, benzoic acid methyl ester, niobe oil, oil of niobe, methyl benzenecarboxylate, essence of niobe, oniobe oil, oxidate le
PubChem CID 7150
ChEBI CHEBI:72775
IUPAC Name methyl benzoate
SMILES COC(=O)C1=CC=CC=C1

Specifications

Flash Point 83.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.09kg
Formula Weight 136.15
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Methyl Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.