Learn More
Medchemexpress LLC Methyl benzyl-L-serinate | 123639-56-5 | 209.25 | 10 G
SDP
Catalog No. 5000449159
Encompass_Preferred
Methyl benzyl-L-serinate is a serine derivative, functioning as a significant chiral building block in organic synthesis and medicinal chemistry. Its structure features a benzyl group on the amino group and a methyl ester at the carboxyl terminus, allowing selective reactions at the serine backbone's hydroxyl group. This strategic protection of functional groups makes it a versatile intermediate for various synthetic pathways.
  • Chiral building block for complex molecules
  • Versatile intermediate for synthetic pathways
  • Spectroscopic data available
  • Utilized in complex molecule synthesis
  • Plays a role in therapeutic agent development
  • Important in organic synthesis
  • Applicable in medicinal chemistry
Catalog No. 50-004-49159 Supplier Medchemexpress LLC Supplier No. HYW003903A10G
Methyl benzyl-L-serinate is a serine derivative, functioning as a significant chiral building block in organic synthesis and medicinal chemistry. Its structure features a benzyl group on the amino group and a methyl ester at the carboxyl terminus, allowing selective reactions at the serine backbone's hydroxyl group. This strategic protection of functional groups makes it a versatile intermediate for various synthetic pathways.
  • Chiral building block for complex molecules
  • Versatile intermediate for synthetic pathways
  • Spectroscopic data available
  • Utilized in complex molecule synthesis
  • Plays a role in therapeutic agent development
  • Important in organic synthesis
  • Applicable in medicinal chemistry