Learn More
MOLECULAR DEPOT LLC Methyl-beta-cyclodextrin-25 mL
Catalog No. 5000013397
Encompass_Preferred
10perc Methyl-beta-cyclodextrin is an aqueous solution of the cyclic methylated dextrin of type Beta This product has been used as versatile tools in molecular assemblies enhancement of solubility and organic synthesis It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only
Catalog No. 50-000-13397 Supplier MOLECULAR DEPOT LLC Supplier No. K1010005C
10perc Methyl-beta-cyclodextrin is an aqueous solution of the cyclic methylated dextrin of type Beta This product has been used as versatile tools in molecular assemblies enhancement of solubility and organic synthesis It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only