Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl-Beta-Cyclodextrin, Spectrum™ Chemical
SDP

Catalog No. 18621122 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
250 g
15 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-122 5 g
18-621-123 10 g
18-621-124 100 g
18-621-125 250 g
18-621-126 15 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-621-122 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M27995GM
Add to Cart
Add to Cart

Synonym: Methyl-B-Cyclodextrin, Beta-Cyclodextrin Methyl Ethers, Methyl Betacyclodextrin

Methyl-beta-cyclodextrin, also known as Cyclodextrins, can form complexes with hydrophobic compounds with use in the food and pharmaceutical industries as a solubilizer, stabilizer, emulsifier and odor reducing agent. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 128446-36-6

Specifications

Quantity 5 g
Percent Purity 98%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Methyl-beta-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.