Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl beta-D-Arabinopyranoside 99.0+%, TCI America™
SDP

Catalog No. M1019100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1019100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1019100MG Supplier TCI America Supplier No. M1019100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reagent for chemical research applications.

Chemical Identifiers

CAS 5328-63-2
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00063261
InChI Key ZBDGHWFPLXXWRD-ARQDHWQXSA-N
PubChem CID 6102754
IUPAC Name (2R,3S,4R,5R)-2-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(CO1)O)O)O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 100 mg
Formula Weight 164.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl beta-D-Arabinopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.