Learn More
Medchemexpress LLC Methyl Blue | 28983-56-4 | MFCD00003507 | >=80% | 5 G
SDP
Catalog No. 5000410804
Encompass_Preferred
Methyl blue belongs to the group of triaminotriphenylmethane dyes. It is extensively used as an antiseptic dye in polychrome staining methods and finds applications in histological and microbiological staining solutions. Methyl blue has also been utilized as a model to investigate the effect of various catalysts on the photodegradation of dyes.
  • Antiseptic dye.
  • Used in polychrome staining methods.
  • Applicable in histological and microbiological staining solutions.
  • Used as a model for studying photodegradation of dyes.
Catalog No. 50-004-10804 Supplier Medchemexpress LLC Supplier No. HYD00035G
Methyl blue belongs to the group of triaminotriphenylmethane dyes. It is extensively used as an antiseptic dye in polychrome staining methods and finds applications in histological and microbiological staining solutions. Methyl blue has also been utilized as a model to investigate the effect of various catalysts on the photodegradation of dyes.
  • Antiseptic dye.
  • Used in polychrome staining methods.
  • Applicable in histological and microbiological staining solutions.
  • Used as a model for studying photodegradation of dyes.