Learn More
Abcam Methyl blue sodium salt, Histology stain for collagen, 50G
Catalog No. 502704771
Encompass_Preferred
MW 799.8 g/mol. Histology stain for collagen in tissue sections. Used in some differential staining techniques such as Mallory′s connective tissue stain and Gomori′s one-step trichrome procedure, and has been incorporated in Sabouraud agar for the identification of Candida albicans strains.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4771 Supplier Abcam Supplier No. AB14627150G
MW 799.8 g/mol. Histology stain for collagen in tissue sections. Used in some differential staining techniques such as Mallory′s connective tissue stain and Gomori′s one-step trichrome procedure, and has been incorporated in Sabouraud agar for the identification of Candida albicans strains.
The product is subject to the following: Abcam Restricted Use Statement