Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl brassylate | 1472-87-3 | MFCD00008464 | 98.0% | 272.39 | 100 G
SDP

Catalog No. 5000455006
Encompass_Preferred

Methyl brassylate (Dimethyl tridecanedioate) is categorized as an ester product. It appears as a white to off-white solid. It is intended for research use only and is not sold to patients. The product is shipped at room temperature within the continental US and should be stored at room temperature for up to 3 years. When stored in solvent, it can be kept at -80°C for 2 years or -20°C for 1 year.

  • Product type: Ester product
  • Appearance: Solid
  • Color: White to off-white
  • Storage conditions: Room temperature (3 years)
  • In solvent: -80°C (2 years), -20°C (1 year)
  • Usage: For research use only

Catalog No. 50-004-55006 Supplier Medchemexpress LLC Supplier No. HYW011637100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl brassylate (Dimethyl tridecanedioate) is categorized as an ester product. It appears as a white to off-white solid. It is intended for research use only and is not sold to patients. The product is shipped at room temperature within the continental US and should be stored at room temperature for up to 3 years. When stored in solvent, it can be kept at -80°C for 2 years or -20°C for 1 year.

  • Product type: Ester product
  • Appearance: Solid
  • Color: White to off-white
  • Storage conditions: Room temperature (3 years)
  • In solvent: -80°C (2 years), -20°C (1 year)
  • Usage: For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.