Learn More
Medchemexpress LLC Methyl brassylate | 1472-87-3 | MFCD00008464 | 98.0% | 272.39 | 500 G
SDP
Catalog No. 5000455036
Encompass_Preferred
Methyl brassylate, also known as Dimethyl tridecanedioate, is an ester product that appears as a white to off-white solid. It is recommended to store this product at room temperature for up to 3 years. When in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year.
  • Ester product
  • White to off-white solid appearance
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year
  • Ships at room temperature in continental US
Catalog No. 50-004-55036 Supplier Medchemexpress LLC Supplier No. HYW011637500G
Methyl brassylate, also known as Dimethyl tridecanedioate, is an ester product that appears as a white to off-white solid. It is recommended to store this product at room temperature for up to 3 years. When in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year.
  • Ester product
  • White to off-white solid appearance
  • Store at room temperature for 3 years
  • Store in solvent at -80°C for 2 years, or -20°C for 1 year
  • Ships at room temperature in continental US