Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl butyrate

Catalog No. 2460935ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-609-35ML 5 mL
24-609-3500ML 500 mL
24-609-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24-609-35ML Supplier Sigma Aldrich Supplier No. 2460935ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl butyrate, one of the odorous compound present in different types of adhesive (hotmelt, vinyl acetate ethylene, starch, polyvinyl acetate and acrylic) used in food packaging, was identified by GC-O-MS (gas chromatography-mass spectrometry-olfactometry).

Specifications

CAS 623-42-7
Density 0.898 g/mL (at 25°C (literature))
Boiling Point 102°C to 103°C (lit.)
Molecular Formula C5H10O2
Refractive Index n20/D 1.385 (literature)
Linear Formula CH3CH2CH2COOCH3
MDL Number MFCD00009391
Quantity 5 mL
Molecular Weight (g/mol) 102.13
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.