Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl butyrate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020190 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0190 1 mL
11-101-8828 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0190 Supplier MilliporeSigma Supelco Supplier No. 193581ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl butyrate is a clear colourless liquid which is slightly soluble in water. It adds flavour and odour to fruits and flowers. It has a characteristic pineapple odour.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 623-42-7
Vapor Pressure 40 mmHg (30°C)
Vapor Density 3.5 (vs. air)
Density 0.898 g/mL (at 25°C)
Flash Point 14°C
Physical Form Neat
Linear Formula CH3 CH2 CH2 COOCH3
MDL Number MFCD00009391
Boiling Point 102°C to 103°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 102.13
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.