Learn More
Medchemexpress LLC Methyl caprooyl tyrosinate | 1104874-94-3 | 98.6% | 293.36 g/mol | C16H23NO4 | 25 MG
SDP
Catalog No. 5000248978
Encompass_Preferred
MHP (methyl caprooyl tyrosinate) is a small-molecule activator of sphingosine kinase 1 (SPHK1) used as a research reagent. It is supplied as a solid (white to off-white) with reported purity of 98.6%, molecular formula C16H23NO4, and CAS 1104874-94-3. The compound is highly soluble in DMSO and can be prepared for in vivo dosing with common co-solvent systems.
Catalog No. 50-002-48978 Supplier Medchemexpress LLC Supplier No. HY10157225MG
MHP (methyl caprooyl tyrosinate) is a small-molecule activator of sphingosine kinase 1 (SPHK1) used as a research reagent. It is supplied as a solid (white to off-white) with reported purity of 98.6%, molecular formula C16H23NO4, and CAS 1104874-94-3. The compound is highly soluble in DMSO and can be prepared for in vivo dosing with common co-solvent systems.