Learn More
Medchemexpress LLC Methyl carbazole-3-carboxylate | 97931-41-4 | MFCD17214817 | 98.1% | 225.24 g/mol | C14H11NO2 | 10 MG
SDP
Catalog No. 5000289137
Encompass_Preferred
Methyl carbazole-3-carboxylate is a carbazole alkaloid derivative used as a research chemical in biochemical and natural-product studies. It is supplied as a light brown to brown solid with reported purity of 98.1%, molecular formula C14H11NO2, and molecular weight 225.24 g/mol. Storage recommendations include room temperature (up to 3 years) or frozen in solvent for long-term storage.
  • Natural product derivative useful for biochemical and natural-product research.
  • High reported purity suitable for analytical and preparative applications.
  • Supplied as a stable solid for ease of handling and storage.
  • Available in small laboratory pack sizes for screening and method development.
  • Documented storage recommendations for standard and solvent conditions.
Catalog No. 50-002-89137 Supplier Medchemexpress LLC Supplier No. HYN328810MG
Methyl carbazole-3-carboxylate is a carbazole alkaloid derivative used as a research chemical in biochemical and natural-product studies. It is supplied as a light brown to brown solid with reported purity of 98.1%, molecular formula C14H11NO2, and molecular weight 225.24 g/mol. Storage recommendations include room temperature (up to 3 years) or frozen in solvent for long-term storage.
  • Natural product derivative useful for biochemical and natural-product research.
  • High reported purity suitable for analytical and preparative applications.
  • Supplied as a stable solid for ease of handling and storage.
  • Available in small laboratory pack sizes for screening and method development.
  • Documented storage recommendations for standard and solvent conditions.