Learn More
Medchemexpress LLC Methyl carnosate | 82684-06-8 | 97.6% | 346.46 | 1 ML
SDP
Catalog No. 5000350741
Encompass_Preferred
Methyl carnosate is a diterpene isolated from *Salvia officinalis* or *Rosmarinus officinalis*. It exhibits potent antioxidant and anti-bacterial activity.
  • Isolated from natural sources like sage and rosemary.
  • Acts as a powerful antioxidant.
  • Demonstrates antibacterial properties.
  • Has cytotoxic effects on certain cancer cell lines such as MIA PaCa-2 and PANC-1.
  • Available in various quantities for research needs.
Catalog No. 50-003-50741 Supplier Medchemexpress LLC Supplier No. HY13615010MM/1ML
Methyl carnosate is a diterpene isolated from *Salvia officinalis* or *Rosmarinus officinalis*. It exhibits potent antioxidant and anti-bacterial activity.
  • Isolated from natural sources like sage and rosemary.
  • Acts as a powerful antioxidant.
  • Demonstrates antibacterial properties.
  • Has cytotoxic effects on certain cancer cell lines such as MIA PaCa-2 and PANC-1.
  • Available in various quantities for research needs.