Learn More
Medchemexpress LLC Methyl carnosate | 82684-06-8 | 97.6% | 346.5 g/mol | C21H30O4 | 5 MG
SDP
Catalog No. 5000222491
Encompass_Preferred
Methyl carnosate is a diterpene natural product isolated from Salvia officinalis and Rosmarinus officinalis, reported to have antioxidant and antibacterial activity. It is supplied for research and analytical applications in solid form and as a 10 mM solution in DMSO. Intended for in vitro use only. CAS 82684-06-8.
  • Natural diterpene with antioxidant and antibacterial activity.
  • High purity (97.62%).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Offered in small research sizes, including 5 mg.
  • Suitable for in vitro biochemical and microbiology studies.
Catalog No. 50-002-22491 Supplier Medchemexpress LLC Supplier No. HY1361505MG
Methyl carnosate is a diterpene natural product isolated from Salvia officinalis and Rosmarinus officinalis, reported to have antioxidant and antibacterial activity. It is supplied for research and analytical applications in solid form and as a 10 mM solution in DMSO. Intended for in vitro use only. CAS 82684-06-8.
  • Natural diterpene with antioxidant and antibacterial activity.
  • High purity (97.62%).
  • Available as a solid and as a 10 mM solution in DMSO.
  • Offered in small research sizes, including 5 mg.
  • Suitable for in vitro biochemical and microbiology studies.