Learn More
Medchemexpress LLC Methyl carnosate | 82684-06-8 | 97.62% | 346.46 | 25 MG
SDP
Catalog No. 5000351140
Encompass_Preferred
Methyl carnosate is a diterpene isolated from Salvia officinalis or Rosmarinus officinalis. It is recognized for its potent antioxidant and anti-bacterial properties. This product is provided as a solid with a high purity of 97.62% and is intended for research use only.
  • Isolated from natural sources like Salvia officinalis or Rosmarinus officinalis
  • Exhibits potent antioxidant activity
  • Demonstrates anti-bacterial activity against organisms such as Bacillus cereus
  • High purity for reliable research applications
Catalog No. 50-003-51140 Supplier Medchemexpress LLC Supplier No. 25MGHY136150
Methyl carnosate is a diterpene isolated from Salvia officinalis or Rosmarinus officinalis. It is recognized for its potent antioxidant and anti-bacterial properties. This product is provided as a solid with a high purity of 97.62% and is intended for research use only.
  • Isolated from natural sources like Salvia officinalis or Rosmarinus officinalis
  • Exhibits potent antioxidant activity
  • Demonstrates anti-bacterial activity against organisms such as Bacillus cereus
  • High purity for reliable research applications