Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl Cedryl Ketone | 32388-55-9 | ≥80.0% | 246.39 | 50 G
SDP

Catalog No. 5000454322
Encompass_Preferred

Methyl Cedryl Ketone is a biochemical reagent suitable for life science research. This organic compound is presented as a liquid, light yellow to yellow in color, with a purity of ≥80.0%.

  • Can be used as a biological material or organic compound for life science related research.
  • Molecular weight of 246.39.
  • Appearance as a liquid with a density of 0.997 g/cm³.
  • Recommended storage conditions vary based on form, including -20°C for 3 years in pure form or -80°C for 6 months in solvent.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-004-54322 Supplier Medchemexpress LLC Supplier No. HYW01340150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl Cedryl Ketone is a biochemical reagent suitable for life science research. This organic compound is presented as a liquid, light yellow to yellow in color, with a purity of ≥80.0%.

  • Can be used as a biological material or organic compound for life science related research.
  • Molecular weight of 246.39.
  • Appearance as a liquid with a density of 0.997 g/cm³.
  • Recommended storage conditions vary based on form, including -20°C for 3 years in pure form or -80°C for 6 months in solvent.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.