Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl cellulose, 9004-67-5, MFCD00081763, 100 g
Catalog No. NC1078521
Encompass
Viscosity: 400 cP, viscosity: 300-560 cP, 2 % in water(20 °C).
Catalog No. NC1078521 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M0262100G
Viscosity: 400 cP, viscosity: 300-560 cP, 2 % in water(20 °C).