Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | 100G
SDP
Catalog No. 5000236569
Encompass_Preferred
Methyl cellulose (viscosity 1500 mPa·s) is a non-ionic cellulose ether supplied as a solid for laboratory and formulation use. It provides thickening, stabilizing, and film-forming properties for research applications in pharmaceuticals, food, cosmetics, and materials. Solubility: DMSO ~4 mg/mL with ultrasonic/warming; product is hygroscopic.
Catalog No. 50-002-36569 Supplier Medchemexpress LLC Supplier No. HY125861C100G
Methyl cellulose (viscosity 1500 mPa·s) is a non-ionic cellulose ether supplied as a solid for laboratory and formulation use. It provides thickening, stabilizing, and film-forming properties for research applications in pharmaceuticals, food, cosmetics, and materials. Solubility: DMSO ~4 mg/mL with ultrasonic/warming; product is hygroscopic.