Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | C6H7O2(OR1)(OR2)(OR3) | 1KG
SDP
Catalog No. 5000234942
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether supplied as a white to off-white solid powder. This viscosity-grade material (nominal 4000 mPa·s) is used as a thickener, gelling agent, stabilizer, and film former in laboratory formulations and industrial processes.
  • Provides reliable thickening and viscosity control.
  • Exhibits thermogelation, forming gels on heating.
  • Water soluble and forms clear, stable gels or films.
  • Consistent physical form and appearance for reproducible formulation performance.
  • Batch testing and certificate of analysis include measured viscosity (example 4466 mPa·s) for quality verification.
  • Offered in practical pack sizes suitable for laboratory use.
Catalog No. 50-002-34942 Supplier Medchemexpress LLC Supplier No. HY125861E1KG
Methyl cellulose is a non-ionic cellulose ether supplied as a white to off-white solid powder. This viscosity-grade material (nominal 4000 mPa·s) is used as a thickener, gelling agent, stabilizer, and film former in laboratory formulations and industrial processes.
  • Provides reliable thickening and viscosity control.
  • Exhibits thermogelation, forming gels on heating.
  • Water soluble and forms clear, stable gels or films.
  • Consistent physical form and appearance for reproducible formulation performance.
  • Batch testing and certificate of analysis include measured viscosity (example 4466 mPa·s) for quality verification.
  • Offered in practical pack sizes suitable for laboratory use.