Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | 10,000-220,000 g/mol | C6H7O2(OR1)(OR2)(OR3) (polymeric anhydroglucose repeating unit) | 5G
SDP
Catalog No. 5000236568
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether supplied as a white, water-soluble powder used as a thickener, binder, film former, and stabilizer in laboratory and formulation applications. It is available in multiple viscosity grades to tailor solution rheology and typically dissolves in cold water to form clear, viscous gels.
  • Non-ionic cellulose ether thickener and gelling agent.
  • Multiple viscosity grades available to tailor rheology.
  • Dissolves in cold water to form clear, viscous gels.
  • Used as a binder, film former, stabilizer, and emulsifier.
  • Supplied as a solid powder suitable for laboratory formulation.
  • Safety considerations: harmful if swallowed; may irritate skin, eyes, and respiratory tract.
Catalog No. 50-002-36568 Supplier Medchemexpress LLC Supplier No. HY1258615G
Methyl cellulose is a non-ionic cellulose ether supplied as a white, water-soluble powder used as a thickener, binder, film former, and stabilizer in laboratory and formulation applications. It is available in multiple viscosity grades to tailor solution rheology and typically dissolves in cold water to form clear, viscous gels.
  • Non-ionic cellulose ether thickener and gelling agent.
  • Multiple viscosity grades available to tailor rheology.
  • Dissolves in cold water to form clear, viscous gels.
  • Used as a binder, film former, stabilizer, and emulsifier.
  • Supplied as a solid powder suitable for laboratory formulation.
  • Safety considerations: harmful if swallowed; may irritate skin, eyes, and respiratory tract.