Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | C20H38O11 | 100G
SDP
Catalog No. 5000239541
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether supplied as a white to off-white solid with a specified viscosity of 4000 mPa·s. It functions as a thickener, stabilizer, film former, and thermogelation agent for use in pharmaceutical formulations, food and cosmetic applications, and laboratory preparations.
  • Provides controllable viscosity and thickening in aqueous systems.
  • Exhibits thermogelation behavior on heating for temperature-responsive gels.
  • Acts as a stabilizer and film former in emulsions and suspensions.
  • Available in multiple viscosity grades and pack sizes for formulation flexibility.
  • White to off-white solid that is easy to handle and store.
Catalog No. 50-002-39541 Supplier Medchemexpress LLC Supplier No. HY125861E100G
Methyl cellulose is a non-ionic cellulose ether supplied as a white to off-white solid with a specified viscosity of 4000 mPa·s. It functions as a thickener, stabilizer, film former, and thermogelation agent for use in pharmaceutical formulations, food and cosmetic applications, and laboratory preparations.
  • Provides controllable viscosity and thickening in aqueous systems.
  • Exhibits thermogelation behavior on heating for temperature-responsive gels.
  • Acts as a stabilizer and film former in emulsions and suspensions.
  • Available in multiple viscosity grades and pack sizes for formulation flexibility.
  • White to off-white solid that is easy to handle and store.