Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | 250G
SDP
Catalog No. 5000235965
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether supplied as a white to off-white powder. It functions as a thickener, stabilizer, emulsifier, and thermogelation agent, characterized by a nominal viscosity of 4000 mPa·s and identified by CAS 9004-67-5. The material is suitable for formulation, laboratory, and pharmaceutical excipient uses and has documented storage stability at room temperature for up to 3 years.
  • Non-ionic cellulose ether for thickening and stabilizing
  • Nominal viscosity 4000 mPa·s for controlled rheology
  • White to off-white solid powder form
  • Suitable for emulsification, film forming, and thermogelation
  • Room temperature shelf life up to 3 years; solvent storage options for extended stability
Catalog No. 50-002-35965 Supplier Medchemexpress LLC Supplier No. HY125861E250G
Methyl cellulose is a non-ionic cellulose ether supplied as a white to off-white powder. It functions as a thickener, stabilizer, emulsifier, and thermogelation agent, characterized by a nominal viscosity of 4000 mPa·s and identified by CAS 9004-67-5. The material is suitable for formulation, laboratory, and pharmaceutical excipient uses and has documented storage stability at room temperature for up to 3 years.
  • Non-ionic cellulose ether for thickening and stabilizing
  • Nominal viscosity 4000 mPa·s for controlled rheology
  • White to off-white solid powder form
  • Suitable for emulsification, film forming, and thermogelation
  • Room temperature shelf life up to 3 years; solvent storage options for extended stability