Learn More
Medchemexpress LLC Methyl cellulose | 9004-67-5 | MFCD00081763 | [C6H7O2(OH)x(OCH3)y]n (x = 1.00-1.55; y = 2.00-1.45; x+y = 3.00) | 50G
SDP
Catalog No. 5000239540
Encompass_Preferred
Methyl cellulose is a non-ionic cellulose ether powder (CAS 9004-67-5) with surface-activity and thermogelation properties. This 1500 mPa·s viscosity grade is supplied as a powder for use as a thickener, stabilizer, emulsifier, and film former in laboratory formulations and product development; supplied in a 50 g pack.
  • Provides viscosity control and thickening in aqueous systems.
  • Exhibits thermogelation, forming gels on heating for temperature-responsive formulations.
  • Acts as a stabilizer and emulsifier for pharmaceuticals, cosmetics, and food research.
  • Water soluble and forms clear gels or films.
  • Suitable for laboratory formulation, rheology testing, and drug delivery studies.
Catalog No. 50-002-39540 Supplier Medchemexpress LLC Supplier No. HY125861C50G
Methyl cellulose is a non-ionic cellulose ether powder (CAS 9004-67-5) with surface-activity and thermogelation properties. This 1500 mPa·s viscosity grade is supplied as a powder for use as a thickener, stabilizer, emulsifier, and film former in laboratory formulations and product development; supplied in a 50 g pack.
  • Provides viscosity control and thickening in aqueous systems.
  • Exhibits thermogelation, forming gels on heating for temperature-responsive formulations.
  • Acts as a stabilizer and emulsifier for pharmaceuticals, cosmetics, and food research.
  • Water soluble and forms clear gels or films.
  • Suitable for laboratory formulation, rheology testing, and drug delivery studies.