Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Methyl Cellulose Solution, 1.5%
SCI_ED

Catalog No. S25428 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
20 mL
Packaging:
1.5%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25428 20 mL 1.5%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25428
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 9004-67-5 , 7732-18-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Quantity 20 mL
Packaging 1.5%
Chemical Name or Material Methyl Cellulose Solution, 1.5%
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.