Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl cellulose, 9004-67-5, MFCD00081763, 100 g
Catalog No. NC1078520
Encompass
Viscosity: 4,000 cP, Methyl cellulose has been used: in colony forming assays, for sperm motility assessment, for the generation of spheroids, and in preparation of agar beads.
Catalog No. NC1078520 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M0512100G
Viscosity: 4,000 cP, Methyl cellulose has been used: in colony forming assays, for sperm motility assessment, for the generation of spheroids, and in preparation of agar beads.