Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cellulose, MP Biomedicals™

Catalog No. MP215549683 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215549683 250 g
ICN15549690 500 g
MP215549691 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215549683 Supplier MP Biomedicals, Inc Supplier No. 0215549683
Add to Cart
Add to Cart

Methyl cellulose , White Powder

  • It is used as a substitute for water-soluble gums,to render paper grease proof in adhesives,as thickening agent in cosmetics,as protective colloid in emulsions,as binder and stabilizer in foods,as bulk producer in the formulation of dietetic foods,Pharmaceutic aid (suspending agent),Laxative.
  • It is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel.
  • It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
MDL Number MFCD00081763
Synonym Cellulose methyl ether, Methocel MC, Cellothyl, Syncelose

Specifications

Color White
pH 6.7
Loss on Drying ≤5.0%
Quantity 250 g
Solubility Information Soluble in cold water. Insoluble in hot water.
InChI Key YLGXILFCIXHCMC-UHFFFAOYNA-N
Molecular Weight (g/mol) 454.51
Physical Form Powder
RTECSNumber FJ5959000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.