Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Cellulose, viscosity of 2% aqueous solution (25deg.C) 1,500cps, MP Biomedicals

Catalog No. p-3572958 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15549583 250 g
ICN15549590 500 g
ICN15549591 1 kg
ICN15549580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15549583 Supplier MP Biomedicals, Inc Supplier No. 0215549583
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
Synonym Cellulose methyl ether
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Quantity 250 g
Chemical Name or Material Methyl Cellulose, Viscosity of 2% aqueous solution at 25°C: 1, 500cps
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.