Learn More
Sigma Aldrich Fine Chemicals Biosciences Methyl cellulose viscosity500G
Catalog No. 502897971
Encompass
Methyl cellulose is a tough non-toxic material. It is odorless and tasteless. Hence methyl cellulose is widely used as an adhesive in the food packaging industry. In water-soluble adhesives it is utilized as a thickening agent. Methyl cellulose is also used as a viscosity control agent. In the cosmetic industry it is used as an emulsifier binder and stabilizer.
Catalog No. 50-289-7971 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M0387500G
Methyl cellulose is a tough non-toxic material. It is odorless and tasteless. Hence methyl cellulose is widely used as an adhesive in the food packaging industry. In water-soluble adhesives it is utilized as a thickening agent. Methyl cellulose is also used as a viscosity control agent. In the cosmetic industry it is used as an emulsifier binder and stabilizer.