Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl chloroformate

Catalog No. M35304100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M35304100G 100 g
M35304500G 500 g
M353045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M35304100G Supplier Sigma Aldrich Supplier No. M35304100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl chloroformate (mcf) is generally used for the derivatization of functional groups such as carboxylic acids, amines, and phenols.mcf can also be used:
  • To activate 3-acylpyridines for nucleophilic addition with alkynyltin reagents to form 2,3-disubstituted 1,2-dihydropyridines
  • Chloroesterification of terminal alkynes to form α-chloro-α,α-unsaturated esters
  • As an electrophilic reagent to mediate the reaction of pyridine with lithium dialkyl- or diarylcuprates to form 4-substituted 1,4-dihydropyridine derivatives
  • To convert nitronates into methoxycarbonyl nitronates

Specifications

CAS 79-22-1
Density 1.223 g/mL (at 25°C (literature))
Boiling Point 70°C to 72°C (lit.)
Molecular Formula C2H3ClO2
Linear Formula ClCOOCH3
Refractive Index n20/D 1.387 (literature)
MDL Number MFCD00000639
Quantity 100 g
Synonym MCF; Chloroformic acid methyl ester
Molecular Weight (g/mol) 94.5
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.