Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Chloroglyoxylate 98.0+%, TCI America™
SDP

Catalog No. C1561250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1561250G 250 g
C156125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1561250G Supplier TCI America Supplier No. C1561250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5781-53-3
Molecular Formula C3H3ClO3
Molecular Weight (g/mol) 122.504
MDL Number MFCD00000705
InChI Key ZXUQEPZWVQIOJE-UHFFFAOYSA-N
Synonym methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
PubChem CID 79846
IUPAC Name methyl 2-chloro-2-oxoacetate
SMILES COC(=O)C(=O)Cl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 250 g
UN Number 2920
Formula Weight 122.50
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl Chloroglyoxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.