Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl Chlorooxoacetate

Catalog No. 1514405G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-144-05G 5 g
15-144-050G 50 g
15-144-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 15-144-05G Supplier Sigma Aldrich Supplier No. 1514405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Regioselective synthesis of fused coumarinsCyclization to form substituted isoxazoles and heterocyclesIntramolecular Wittig reactionsSilyl enol ether acylationIron-mediated cleavage of C-C bonds

Specifications

CAS 5781-53-3
Density 1.332 g/mL at 25°C
Boiling Point 118°C to 120°C
Molecular Formula C3H3ClO3
Refractive Index n20/D 1.419
Linear Formula ClCOCOOCH3
MDL Number MFCD00000705
Quantity 5 g
Synonym Monomethyl oxalyl chloride; Chloroglyoxylic acid methyl ester; Methyl (chloroformyl)formate; Methyl 2-chloro-2-oxoacetate; Methyl chloroglyoxalate; Methyl oxalyl chloride
Molecular Weight (g/mol) 122.51 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.