Learn More
Medchemexpress LLC HY-107830 100mg Medchemexpress, Methyl cholate CAS:1448-36-8 Purity:>98%
SDP
Catalog No. 501968474
Encompass_Preferred
Medchemexpress, HY-107830 100mg Methyl cholate CAS:1448-36-8 Methyl Cholate is methyl ester form of Cholic acid. Cholic acid is one of the major bile acids produced by the liver, where it is synthesized from cholesterol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8474 Supplier Medchemexpress LLC Supplier No. HY107830100MG
Medchemexpress, HY-107830 100mg Methyl cholate CAS:1448-36-8 Methyl Cholate is methyl ester form of Cholic acid. Cholic acid is one of the major bile acids produced by the liver, where it is synthesized from cholesterol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.