Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl cholate | 1448-36-8 | MFCD00064934 | 99.8% | 422.6 g/mol | C25H42O5 | 1 ML
SDP

Catalog No. 5000242761
Encompass_Preferred

Methyl cholate is the methyl ester of cholic acid provided as a high-purity biochemical reagent for research and analytical applications. It is offered both as a solid and as ready-to-use DMSO solutions, and is commonly used in studies of bile acids, supramolecular carriers, and sterol ester chemistry.

  • High purity (99.8%).
  • Chemical formula C25H42O5; molecular weight 422.6 g/mol.
  • Available as 10 mM solution in DMSO or in solid pack sizes.
  • Cas registry number 1448-36-8 for unambiguous identification.
  • Suitable for research use and analytical standards.

Catalog No. 50-002-42761 Supplier Medchemexpress LLC Supplier No. HY10783010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl cholate is the methyl ester of cholic acid provided as a high-purity biochemical reagent for research and analytical applications. It is offered both as a solid and as ready-to-use DMSO solutions, and is commonly used in studies of bile acids, supramolecular carriers, and sterol ester chemistry.

  • High purity (99.8%).
  • Chemical formula C25H42O5; molecular weight 422.6 g/mol.
  • Available as 10 mM solution in DMSO or in solid pack sizes.
  • Cas registry number 1448-36-8 for unambiguous identification.
  • Suitable for research use and analytical standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.