Learn More
Medchemexpress LLC Methyl cinnamate | 103-26-4 | MFCD00008458 | 99.6% | 162.19 g/mol | C10H10O2 | 1 ML
SDP
Catalog No. 5000300566
Encompass_Preferred
Methyl cinnamate is methyl 3-phenylpropenoate supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is a naturally occurring flavor compound with reported tyrosinase inhibitory and antimicrobial activities, commonly used as a reference in biochemical and food science research. The solution provides a convenient, accurately prepared stock for in vitro assays and analytical applications.
  • Ready-to-use 10 mM solution in DMSO for immediate use
  • Accurately prepared 1 mL vial to simplify assay setup
  • High purity for reproducible experimental results
  • Suitable as a tyrosinase inhibitor and antimicrobial reference
  • Stable when stored under recommended conditions
Catalog No. 50-003-00566 Supplier Medchemexpress LLC Supplier No. HYW01721210MM/1ML
Methyl cinnamate is methyl 3-phenylpropenoate supplied as a ready-to-use 10 mM solution in DMSO (1 mL). It is a naturally occurring flavor compound with reported tyrosinase inhibitory and antimicrobial activities, commonly used as a reference in biochemical and food science research. The solution provides a convenient, accurately prepared stock for in vitro assays and analytical applications.
  • Ready-to-use 10 mM solution in DMSO for immediate use
  • Accurately prepared 1 mL vial to simplify assay setup
  • High purity for reproducible experimental results
  • Suitable as a tyrosinase inhibitor and antimicrobial reference
  • Stable when stored under recommended conditions