Learn More
Medchemexpress LLC Methyl cinnamate | 103-26-4 | MFCD00008458 | 99.6% | 162.19 g/mol | C10H10O2 | 10 G
SDP
Catalog No. 5000300153
Encompass_Preferred
Methyl cinnamate is an aromatic ester (methyl 3-phenylpropenoate) commonly used as a reagent in research applications and as a flavor and fragrance component. The material is characterized for analytical use and is supplied with supporting documentation for laboratory handling and quality verification.
  • Used as an aromatic ester reagent and flavor and fragrance component.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • High purity (99.6%).
  • Molecular formula C10H10O2; molecular weight 162.19 g/mol.
  • Available in small laboratory pack sizes suitable for analytical work.
Catalog No. 50-003-00153 Supplier Medchemexpress LLC Supplier No. HYW01721210G
Methyl cinnamate is an aromatic ester (methyl 3-phenylpropenoate) commonly used as a reagent in research applications and as a flavor and fragrance component. The material is characterized for analytical use and is supplied with supporting documentation for laboratory handling and quality verification.
  • Used as an aromatic ester reagent and flavor and fragrance component.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • High purity (99.6%).
  • Molecular formula C10H10O2; molecular weight 162.19 g/mol.
  • Available in small laboratory pack sizes suitable for analytical work.