Learn More
Medchemexpress LLC Methyl cinnamate | 103-26-4 | MFCD00008458 | 99.6% | 162.19 g·mol⁻¹ | C10H10O2 | 25 G
SDP
Catalog No. 5000304103
Encompass_Preferred
Methyl cinnamate (CAS 103-26-4) is an aromatic ester used as a flavor and fragrance component and as a reagent in organic synthesis and biological assays. Supplied as a high-purity crystalline compound, it is suitable for analytical, preparative, and research applications.
  • High purity for analytical use.
  • Molecular formula C10H10O2; molecular weight 162.19 g·mol⁻¹.
  • Common applications include flavor and fragrance studies and small-molecule research.
  • Available in multiple laboratory pack sizes for convenience.
  • Stable under standard laboratory storage conditions and easy to handle.
Catalog No. 50-003-04103 Supplier Medchemexpress LLC Supplier No. 25GHYW017212
Methyl cinnamate (CAS 103-26-4) is an aromatic ester used as a flavor and fragrance component and as a reagent in organic synthesis and biological assays. Supplied as a high-purity crystalline compound, it is suitable for analytical, preparative, and research applications.
  • High purity for analytical use.
  • Molecular formula C10H10O2; molecular weight 162.19 g·mol⁻¹.
  • Common applications include flavor and fragrance studies and small-molecule research.
  • Available in multiple laboratory pack sizes for convenience.
  • Stable under standard laboratory storage conditions and easy to handle.