Learn More
Selleck Chemical LLC Methyl cinnamate
SDP
Catalog No. 5000003766
Encompass_Preferred
Methyl cinnamate an active component of Zanthoxylum armatum is a widely used natural flavor compound with antimicrobial and tyrosinase inhibitor activities Methyl Cinnamate Inhibits Adipocyte Differentiation via Activation of the CaMKK2--AMPK Pathway in 3T3-L1 Preadipocytes
Catalog No. 50-000-03766 Supplier Selleck Chemical LLC Supplier No. S622125MG
Methyl cinnamate an active component of Zanthoxylum armatum is a widely used natural flavor compound with antimicrobial and tyrosinase inhibitor activities Methyl Cinnamate Inhibits Adipocyte Differentiation via Activation of the CaMKK2--AMPK Pathway in 3T3-L1 Preadipocytes