Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cis-4-Aminocyclohexanecarboxylate Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M19855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19855G 5 g
M19851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19855G Supplier TCI America Supplier No. M19855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 61367-16-6 | C8H16ClNO2 | 193.671 g/mol

  • Purity: ≥98.0% (Nonaqueous Titration, Argentometric Titration)
  • Physical Form: White to almost white powder to crystal
  • Melting Point: 188°C
  • Molecular Formula: C8H15NO2·HCl
  • Molecular Weight: 193.67
  • Storage Temperature: Room temperature (recommended in a cool and dark place, <15°C)
  • Structure Analysis: Confirmed by NMR

Chemical Identifiers

CAS 61367-16-6
Molecular Formula C8H16ClNO2
Molecular Weight (g/mol) 193.671
MDL Number MFCD08274537
InChI Key NHAYDXCUCXRAMF-UHFFFAOYSA-N
Synonym cis-4-Aminocyclohexanecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 18521575
IUPAC Name methyl 4-aminocyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)N.Cl

Specifications

Melting Point 188°C
Color White
Quantity 5 g
Formula Weight 193.67
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl cis-4-Aminocyclohexanecarboxylate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.