Learn More
Medchemexpress LLC Methyl citrate | 26163-61-1 | 98.0% | 206.15 | 25 MG
SDP
Catalog No. 5000426749
Encompass_Preferred
Methyl citrate is a monoamine oxidase B (MAO-B) inhibitor isolated from the fruits of *Opuntia ficus-indica var. saboten Makino*.
  • Inhibits monoamine oxidase B
  • Has an IC50 of 0.23 mM
  • Isolated from *Opuntia ficus-indica var. saboten Makino* fruits
Catalog No. 50-004-26749 Supplier Medchemexpress LLC Supplier No. 25MGHYN9540
Add to Cart
Add to Cart
Methyl citrate is a monoamine oxidase B (MAO-B) inhibitor isolated from the fruits of *Opuntia ficus-indica var. saboten Makino*.
  • Inhibits monoamine oxidase B
  • Has an IC50 of 0.23 mM
  • Isolated from *Opuntia ficus-indica var. saboten Makino* fruits