Learn More
Medchemexpress LLC Methyl clofenapate | 21340-68-1 | 99.0% | 304.77 | 25 MG
SDP
Catalog No. 5000393276
Encompass_Preferred
Methyl clofenapate is a chemical compound supplied as a white to off-white solid. It is primarily used as a laboratory chemical for research purposes and the manufacture of substances, exhibiting good stability under recommended storage conditions.
  • For research use only
  • Stable under recommended storage conditions
  • Supplied as a solid (white to off-white)
  • High purity (99.04%)
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years
Catalog No. 50-003-93276 Supplier Medchemexpress LLC Supplier No. 25MGHY164082
Methyl clofenapate is a chemical compound supplied as a white to off-white solid. It is primarily used as a laboratory chemical for research purposes and the manufacture of substances, exhibiting good stability under recommended storage conditions.
  • For research use only
  • Stable under recommended storage conditions
  • Supplied as a solid (white to off-white)
  • High purity (99.04%)
  • Recommended storage for powder: -20°C for 3 years; 4°C for 2 years