Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl crotonate, 96%, trans-isomer

Catalog No. p-7022536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1536322 100 g
AAA1536336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1536322 Supplier Thermo Scientific Chemicals Supplier No. A1536322
Only null left
Add to Cart

CAS: 623-43-8 | C5H8O2 | 100.12 g/mol

Methyl crotonate was used to investigate chemo selectivity in the reaction between methyl crotonate and benzyl amine catalyzed by lipase B from Candida Antarctica using solvent engineering. It was used as starting reagent during the total synthesis of phytotoxins solanapyrones D(1) and E(2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl crotonate was used to investigate chemo selectivity in the reaction between methyl crotonate and benzyl amine catalyzed by lipase B from Candida Antarctica using solvent engineering. It was used as starting reagent during the total synthesis of phytotoxins solanapyrones D(1) and E(2).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-43-8
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009287
InChI Key MCVVUJPXSBQTRZ-ONEGZZNKSA-N
Synonym methyl crotonate, e-methyl but-2-enoate, trans-methyl crotonate, methyl e-crotonate, methyl 2-butenoate, methyl trans-crotonate, 2-butenoic acid, methyl ester, 2e, trans-2-butenoic acid methyl ester, methyl trans-2-butenoate, e-crotonic acid methyl ester
PubChem CID 638132
IUPAC Name methyl (E)-but-2-enoate
SMILES COC(=O)\C=C\C

Specifications

Density 0.946
Boiling Point 118°C to 120°C
Flash Point 4°C (40°F)
Refractive Index 1.4255
Quantity 100 g
UN Number UN3272
Beilstein 1720292
Merck Index 14,2597
Solubility Information Insoluble in water.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material Methyl crotonate
Show More Show Less
Hazard Category H225-H319
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 210-793-7
RTECSNumber GQ5710000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.