Learn More
Apexbio Technology LLC Methyl-β-cyclodextrin 128446-36-6 500mg
SDP
Catalog No. 5000094713
Encompass_Preferred
Methyl-beta-cyclodextrin (128446-36-6) is a cyclic oligosaccharide derivative used to extract cholesterol from cellular membranes thereby disrupting cholesterol-dependent membrane processes Methyl-beta-cyclodextrin exerts its biological activity primarily through depletion of membrane cholesterol which interferes with clathrin-mediated endocytosis membrane signaling and the formation of cell adhesion and migration structures Based on these pharmacological properties methyl-beta-cyclodextrin holds research potential in studies of endocytosis signal transduction membrane dynamics and cell motility
Catalog No. 50-000-94713 Supplier Apexbio Technology LLC Supplier No. C6939500
Methyl-beta-cyclodextrin (128446-36-6) is a cyclic oligosaccharide derivative used to extract cholesterol from cellular membranes thereby disrupting cholesterol-dependent membrane processes Methyl-beta-cyclodextrin exerts its biological activity primarily through depletion of membrane cholesterol which interferes with clathrin-mediated endocytosis membrane signaling and the formation of cell adhesion and migration structures Based on these pharmacological properties methyl-beta-cyclodextrin holds research potential in studies of endocytosis signal transduction membrane dynamics and cell motility