Learn More
Medchemexpress LLC Methyl-β-cyclodextrin | 128446-36-6 | MFCD00074980 | 100.0% | 1310 g/mol | C54H94O35 | 25 G
SDP
Catalog No. 5000249096
Encompass_Preferred
Methyl-β-cyclodextrin is a randomly methylated derivative of β-cyclodextrin (a cyclic heptasaccharide) used to increase the aqueous solubility of hydrophobic compounds and to extract cholesterol from biological membranes. It is supplied as a white to off-white powder for biochemical and cell biology research.
  • Solubilizes hydrophobic compounds for assays and formulations.
  • Depletes cholesterol from cell membranes to study membrane biology.
  • Water-soluble powder with an average molecular weight around 1,310 g/mol.
  • High reagent purity suitable for biochemical and cell culture applications.
  • Available in research pack sizes commonly including 25 g.
Catalog No. 50-002-49096 Supplier Medchemexpress LLC Supplier No. HY10146125G
Methyl-β-cyclodextrin is a randomly methylated derivative of β-cyclodextrin (a cyclic heptasaccharide) used to increase the aqueous solubility of hydrophobic compounds and to extract cholesterol from biological membranes. It is supplied as a white to off-white powder for biochemical and cell biology research.
  • Solubilizes hydrophobic compounds for assays and formulations.
  • Depletes cholesterol from cell membranes to study membrane biology.
  • Water-soluble powder with an average molecular weight around 1,310 g/mol.
  • High reagent purity suitable for biochemical and cell culture applications.
  • Available in research pack sizes commonly including 25 g.