Learn More
Medchemexpress LLC Methyl-β-cyclodextrin | 128446-36-6 | MFCD00074980 | 99.9% | 1303.3 g/mol | C54H94O35 | 100 G
SDP
Catalog No. 5000249068
Encompass_Preferred
Methyl-β-cyclodextrin is a methylated derivative of β-cyclodextrin used to solubilize hydrophobic compounds and to deplete membrane cholesterol in biological research. It forms inclusion complexes with nonpolar molecules, improving aqueous solubility and facilitating delivery and uptake in cellular assays and formulations.
  • Enhances solubility of hydrophobic molecules for biological and formulation studies.
  • Depletes membrane cholesterol to enable mechanistic studies of membrane-dependent processes.
  • Facilitates delivery of poorly soluble compounds in aqueous media.
  • Modulates endocytosis pathways in cell-based assays.
  • High purity supports reproducible experimental results.
  • Available in multiple pack sizes to suit lab-scale needs.
Catalog No. 50-002-49068 Supplier Medchemexpress LLC Supplier No. HY101461100G
Methyl-β-cyclodextrin is a methylated derivative of β-cyclodextrin used to solubilize hydrophobic compounds and to deplete membrane cholesterol in biological research. It forms inclusion complexes with nonpolar molecules, improving aqueous solubility and facilitating delivery and uptake in cellular assays and formulations.
  • Enhances solubility of hydrophobic molecules for biological and formulation studies.
  • Depletes membrane cholesterol to enable mechanistic studies of membrane-dependent processes.
  • Facilitates delivery of poorly soluble compounds in aqueous media.
  • Modulates endocytosis pathways in cell-based assays.
  • High purity supports reproducible experimental results.
  • Available in multiple pack sizes to suit lab-scale needs.