Learn More
Medchemexpress LLC HY-101461 1g Medchemexpress, Methyl-β-cyclodextrin CAS:128446-36-6 Purity:>98%
SDP
Catalog No. 501932140
Encompass_Preferred
Medchemexpress, HY-101461 1g Methyl-β-cyclodextrin CAS:128446-36-6 Methyl-β-cyclodextrin, a cyclic heptasaccharide, is an effective agent for the depletion of cholesterol from cells; also inhibits PEL cell growth with an IC 50 of 3.33-4.23 mM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2140 Supplier Medchemexpress LLC Supplier No. HY1014611G
Medchemexpress, HY-101461 1g Methyl-β-cyclodextrin CAS:128446-36-6 Methyl-β-cyclodextrin, a cyclic heptasaccharide, is an effective agent for the depletion of cholesterol from cells; also inhibits PEL cell growth with an IC 50 of 3.33-4.23 mM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.