Learn More
Medchemexpress LLC Methyl cyclohexanecarboxylate | 4630-82-4 | 142.20 | 5 G
SDP
Catalog No. 5000446415
Encompass_Preferred
Methyl cyclohexanecarboxylate is an endogenous metabolite.
  • Intended for research use only
  • Colorless to light yellow liquid appearance
  • Soluble in DMSO (requires ultrasonic treatment)
  • Available in various sizes
  • Can be stored at -20°C for 3 years or 4°C for 2 years in pure form
  • Stock solutions in solvent should be aliquoted to prevent freeze-thaw inactivation
  • Key applications include natural products disease research and metabolic studies
Catalog No. 50-004-46415 Supplier Medchemexpress LLC Supplier No. HYW0077045G
Methyl cyclohexanecarboxylate is an endogenous metabolite.
  • Intended for research use only
  • Colorless to light yellow liquid appearance
  • Soluble in DMSO (requires ultrasonic treatment)
  • Available in various sizes
  • Can be stored at -20°C for 3 years or 4°C for 2 years in pure form
  • Stock solutions in solvent should be aliquoted to prevent freeze-thaw inactivation
  • Key applications include natural products disease research and metabolic studies