Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl cyclopentanecarboxylate, 97%

Catalog No. AC450920250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450920250 25 g
AC450920050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450920250 Supplier Thermo Scientific Chemicals Supplier No. 450920250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4630-80-2 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4630-80-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00019290
InChI Key IIHIJFJSXPDTNO-UHFFFAOYSA-N
Synonym cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
PubChem CID 78365
IUPAC Name methyl cyclopentanecarboxylate
SMILES COC(=O)C1CCCC1

Specifications

Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material Methyl cyclopentanecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a well-ventilated place. Keep container tightly closed.
EINECSNumber 225-049-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.