Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl β-D-Galactopyranoside | 1824-94-8 | 99.9% | 100 MG
SDP

Catalog No. 5000342320
Encompass_Preferred

Methyl β-D-Galactopyranoside is an endogenous metabolite and a chemical reagent primarily used in industrial and scientific research. This high-purity compound presents as a white to off-white solid and requires specific storage conditions to maintain its integrity. It is important to handle this substance with care as it can cause skin and serious eye irritation, and may also lead to respiratory irritation.

  • High purity: 99.9%
  • CAS Number: 1824-94-8
  • Appearance: White to off-white solid
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Causes skin and serious eye irritation
  • May cause respiratory irritation

Catalog No. 50-003-42320 Supplier Medchemexpress LLC Supplier No. HY128737100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl β-D-Galactopyranoside is an endogenous metabolite and a chemical reagent primarily used in industrial and scientific research. This high-purity compound presents as a white to off-white solid and requires specific storage conditions to maintain its integrity. It is important to handle this substance with care as it can cause skin and serious eye irritation, and may also lead to respiratory irritation.

  • High purity: 99.9%
  • CAS Number: 1824-94-8
  • Appearance: White to off-white solid
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Causes skin and serious eye irritation
  • May cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.