Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl α-D-galactopyranoside | 3396-99-4 | 194.18 g/mol | C7H14O6 | 25 G
SDP

Catalog No. 5000209197
Encompass_Preferred

Methyl α-D-galactopyranoside is a methyl glycoside of D-galactose used as a carbohydrate reagent in biochemical assays and carbohydrate chemistry.

  • Used as a reagent in biochemical and enzymatic assays
  • Serves as a methyl glycoside derivative of D-galactose for structure-function studies
  • Molecular formula C7H14O6, molecular weight 194.18 g/mol
  • CAS number 3396-99-4 for unambiguous identification
  • Supplied in laboratory-scale packs suitable for research use

Catalog No. 50-002-09197 Supplier Medchemexpress LLC Supplier No. 25GHYW088238
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Methyl α-D-galactopyranoside is a methyl glycoside of D-galactose used as a carbohydrate reagent in biochemical assays and carbohydrate chemistry.

  • Used as a reagent in biochemical and enzymatic assays
  • Serves as a methyl glycoside derivative of D-galactose for structure-function studies
  • Molecular formula C7H14O6, molecular weight 194.18 g/mol
  • CAS number 3396-99-4 for unambiguous identification
  • Supplied in laboratory-scale packs suitable for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.